Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207131.1 | NP_997014.1 | 1 | 374 | ||
GenBank mRNA | AK143561.1 | - | 1 | 375 | ||
AY570294.1 | - | 1 | 374 | |||
BC108953.1 | - | 1 | 311 | 40045189 | ||
BC108954.1 | - | 1 | 311 | 40045190 |