Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310332.1 | NP_001297261.1 | 11 | 1914 | ||
NM_017999.4 | NP_060469.4 | 11 | 2258 | |||
GenBank mRNA | AB265810.1 | - | 11 | 2009 | ||
AK000973.1 | - | 4 | 534 | |||
AK027154.1 | - | 3 | 651 | |||
AK055542.1 | - | 5 | 1055 | |||
AK122711.1 | - | 10 | 1543 | |||
AK225832.1 | - | 11 | 2208 | |||
AK291247.1 | - | 11 | 2210 | |||
AY256461.1 | - | 11 | 1902 | |||
BC009821.2 | - | 4 | 612 | |||
BC012077.1 | - | 11 | 2178 | 4653017 | ||
BC017376.2 | - | 4 | 602 |