Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014442.2 | NP_001014442.2 | 4 | 332 | ||
NM_001135816.1 | NP_001129288.1 | 4 | 229 | |||
GenBank mRNA | BC015452.1 | - | 4 | 334 | ||
BC073902.1 | - | 4 | 332 | |||
BC132842.1 | - | 2 | 168 | |||
BC132846.1 | - | 2 | 168 | |||
BC142999.1 | - | 4 | 229 |