Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017895.7 | NP_060365.7 | 3, 4 | 452 | ||
XM_011528888.2 | XP_011527190.1 | 3, 4 | 423 | |||
GenBank mRNA | AK022979.1 | - | 3, 4 | 322 | ||
AK300135.1 | - | 3, 4 | 331 | |||
AL832131.1 | - | 3, 4 | 368 | |||
AY044431.1 | - | 3, 4 | 391 | |||
BC009304.1 | - | 3, 4 | 310 | |||
BC011927.2 | - | 3, 4 | 306 | |||
BC016060.1 | - | 3, 4 | 306 | |||
BC126287.1 | - | 3, 4 | 441 | 8992003 | ||
BC130275.1 | - | 3, 4 | 441 | 40146306 | ||
BC144125.1 | - | 3, 4 | 441 | 9052645 | ||
BF026095.1 | - | 3, 4 | 400 | |||
DB442175.2 | - | 3, 4 | 454 | |||
EU718675.1 | - | 3, 4 | 359 |