Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 12 | 1613 | ||
NM_031921.5 | NP_114127.3 | 14 | 1496 | |||
XM_005244806.3 | XP_005244863.1 | 14 | 1519 | |||
XM_011542241.2 | XP_011540543.1 | 15 | 1654 | |||
XM_011542243.2 | XP_011540545.1 | 13 | 1232 | |||
XM_011542244.1 | XP_011540546.1 | 14 | 1663 | |||
GenBank mRNA | AB033099.1 | - | 12 | 1702 | ||
AF343078.1 | - | 14 | 1429 | |||
AK024028.1 | - | 12 | 1081 | |||
AK127857.1 | - | 8 | 2267 | |||
AK290586.1 | - | 14 | 1488 | |||
AL834179.1 | - | 4 | 2264 | |||
BC002542.2 | - | 14 | 1422 | 3138578 | ||
BC018701.1 | - | 14 | 1422 | 3139886 |