Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000618.4 | NP_000609.1 | 1, 2 | 335 | ||
NM_001111283.2 | NP_001104753.1 | 1, 2 | 335 | |||
XM_017019260.1 | XP_016874749.1 | 1, 2 | 364 | |||
GenBank mRNA | AB209184.1 | - | 1, 2 | 286 | ||
AK312231.1 | - | 1, 2 | 234 | |||
AL603526.1 | - | 1, 2 | 302 | |||
AL710591.1 | - | 1, 2 | 336 | |||
BC148266.1 | - | 1, 2 | 93 | 8860206 | ||
BC152321.1 | - | 1, 2 | 216 | 40129266 | ||
CR541861.1 | - | 1, 2 | 71 | |||
M11568.1 | - | 1, 2 | 253 | |||
M27544.1 | - | 1, 2 | 219 | |||
M29644.1 | - | 1, 2 | 82 | |||
U40870.1 | - | 1 | 14 | |||
X00173.1 | - | 1, 2 | 82 | |||
X56773.1 | - | 1, 2 | 71 | |||
X56774.1 | - | 1, 2 | 71 | |||
X57025.1 | - | 1, 2 | 237 |