Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009573.2 | NP_001009573.2 | 7, 8 | 677 | ||
GenBank mRNA | BC141289.1 | - | 6, 7 | 589 | 9055901 | |
EF127645.1 | - | 6, 7 | 625 |