Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009959.2 | NP_001009959.1 | 2 | 477 | ||
NM_001304344.1 | NP_001291273.1 | 2 | 325 | |||
NM_001304345.1 | NP_001291274.1 | 2 | 337 | |||
NM_001304346.1 | NP_001291275.1 | 1 | 510 | |||
NM_020711.2 | NP_065762.1 | 1 | 510 | |||
GenBank mRNA | AB033015.1 | - | 1 | 252 | ||
AK295844.1 | - | 1 | 386 | |||
AV727436.1 | - | 2 | 398 | |||
BC026345.1 | - | 1 | 336 | 4798869 | ||
BI598234.1 | - | 1 | 466 | |||
DA248817.1 | - | 2 | 337 | |||
DA288720.1 | - | 2 | 264 | |||
DC322056.1 | - | 2 | 478 | |||
DC331726.1 | - | 1 | 511 | |||
DC343891.1 | - | 1 | 496 | |||
DQ334271.1 | - | 1 | 333 |