Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012310.2 | NP_001012310.2 | 6 | 1246 | ||
GenBank mRNA | AK149369.1 | - | 6 | 1252 | ||
BC035527.1 | - | 5 | 1567 | |||
BC042739.1 | - | 5 | 1374 | |||
BC055027.1 | - | 6 | 1239 | |||
BC069939.1 | - | 6 | 1235 | 5053384 | ||
BC089616.1 | - | 6 | 1246 | 6760348 |