Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014976.2 | NP_001014976.1 | 3 | 821 | ||
GenBank mRNA | AK129072.1 | - | 3 | 788 | ||
AY588413.1 | - | 3 | 759 | |||
BC082603.1 | - | 3 | 821 | 30668743 | ||
BC145844.1 | - | 3 | 792 | 40131147 | ||
BC145846.1 | - | 3 | 738 | 40131149 |