Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022263.2 | NP_071599.2 | 1 | 350 | ||
GenBank mRNA | AF271155.1 | - | 1 | 46 | ||
BC079062.1 | - | 1 | 350 | 7115934 |