Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023072.2 | NP_075560.2 | 2 | 417 | ||
XM_005260023.3 | XP_005260080.1 | 2 | 228 | |||
XM_017027153.1 | XP_016882642.1 | 2 | 451 | |||
XM_017027154.1 | XP_016882643.1 | 2 | 452 | |||
XM_017027155.1 | XP_016882644.1 | 2 | 452 | |||
XM_017027157.1 | XP_016882646.1 | 2 | 453 | |||
GenBank mRNA | AK024452.1 | - | 2 | 417 |