Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199187.2 | NP_001186116.1 | 2, 3 | 215 | ||
NM_026937.3 | NP_081213.1 | 2, 3 | 228 | |||
GenBank mRNA | AK007519.1 | - | 2, 3 | 212 | ||
AK053811.1 | - | 2, 3 | 229 | |||
AK139944.1 | - | 2, 3 | 215 | |||
AK144614.1 | - | 2, 3 | 189 | |||
AK171958.1 | - | 2, 3 | 189 | |||
BB844213.1 | - | 2, 3 | 209 | |||
BC030905.1 | - | 2, 3 | 194 | 4503445 |