Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027644.1 | NP_081920.1 | 5 | 403 | ||
GenBank mRNA | AB049453.1 | - | 5 | 403 | ||
AK016509.1 | - | 5 | 381 | |||
AK049297.1 | - | 5 | 420 | |||
BC145586.1 | - | 5 | 419 | |||
BC150797.1 | - | 5 | 419 | 9087708 |