Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039081.2 | NP_001034170.1 | 13 | 1631 | ||
NM_001286221.1 | NP_001273150.1 | 15 | 1762 | |||
NM_001286222.1 | NP_001273151.1 | 15 | 1816 | |||
NM_001286223.1 | NP_001273152.1 | 13 | 1612 | |||
NM_027696.3 | NP_081972.2 | 15 | 1819 | |||
GenBank mRNA | AK016915.1 | - | 7 | 1061 | ||
AK043136.1 | - | 2 | 1550 | |||
AK129405.1 | - | 15 | 1739 | |||
AK136467.1 | - | 3 | 475 | |||
AY769435.1 | - | 14 | 1611 | |||
AY769436.1 | - | 13 | 1534 | |||
AY769437.1 | - | 13 | 1584 | |||
AY769438.1 | - | 15 | 1684 | |||
BC108273.1 | - | 13 | 1598 | 30080243 | ||
BC130227.1 | - | 13 | 1583 | 40141863 |