Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293806.1 | NP_001280735.1 | 23 | 4061 | ||
NM_027777.2 | NP_082053.1 | 22 | 3941 | |||
GenBank mRNA | AK017309.1 | - | 10 | 1358 | ||
BC090845.1 | - | 22 | 3941 | 6400629 |