Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027889.1 | NP_082165.1 | 15, 16 | 2708 | ||
GenBank mRNA | AK004695.1 | - | 15, 16 | 2683 | ||
AK037658.1 | - | 13, 14 | 3163 | |||
AK038682.1 | - | 2, 3 | 341 | |||
AK090030.1 | - | 15, 16 | 2689 | |||
AK153963.1 | - | 15, 16 | 2681 | |||
AK162897.1 | - | 16, 17 | 2728 | |||
AK210514.1 | - | 1, 2 | 56 | |||
BC016258.1 | - | 15, 16 | 2677 | |||
BC029004.1 | - | 15, 16 | 2708 | 5360111 |