Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286468.1 | NP_001273397.1 | 7 | 1228 | ||
NM_029270.2 | NP_083546.2 | 9 | 1830 | |||
NM_146161.3 | NP_666273.1 | 8 | 1636 | |||
GenBank mRNA | AK080986.1 | - | 9 | 1830 | ||
AK134817.1 | - | 7 | 1452 | |||
BC023344.1 | - | 7 | 1242 | 5064885 | ||
BC025502.1 | - | 8 | 1485 | 5321985 | ||
BC027070.1 | - | 6 | 1074 | 5358648 |