Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007090.2 | NP_001007091.1 | 1 | 641 | ||
GenBank mRNA | AF394908.1 | - | 1 | 494 | ||
AK058131.1 | - | 1 | 641 | |||
AK058156.1 | - | 1 | 21 | |||
BC031245.1 | - | 1 | 589 | 5266320 |