Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008385.1 | NP_001008386.1 | 3 | 276 | ||
GenBank mRNA | AY788892.1 | - | 3 | 276 |