Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010875.3 | NP_001010875.1 | 3 | 328 | ||
NM_001286806.1 | NP_001273735.1 | 2 | 209 | |||
XM_005266321.2 | XP_005266378.1 | 3 | 254 | |||
XM_005266322.1 | XP_005266379.1 | 3 | 254 | |||
XM_006719793.3 | XP_006719856.1 | 3 | 500 | |||
XM_006719794.1 | XP_006719857.1 | 3 | 378 | |||
XM_017020523.1 | XP_016876012.1 | 3 | 325 | |||
XM_017020524.1 | XP_016876013.1 | 3 | 378 | |||
GenBank mRNA | AK094133.1 | - | 3 | 255 | ||
AK302967.1 | - | 2 | 136 | |||
BC132739.1 | - | 3 | 251 | 40146761 | ||
BC136760.1 | - | 3 | 251 | 9020750 | ||
DC322809.1 | - | 3 | 285 |