Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010893.2 | NP_001010893.1 | 1 | 333 | ||
GenBank mRNA | AK095808.1 | - | 1 | 1538 | ||
AY825924.1 | - | 1 | 333 | |||
BC136625.1 | - | 1 | 413 | 9020615 | ||
BC136631.1 | - | 1 | 413 | 9020621 |