Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011980.1 | NP_001011980.1 | 9 | 972 | ||
GenBank mRNA | BC089842.1 | - | 9 | 972 | 7366262 |