Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012056.1 | NP_001012056.1 | 3 | 441 | ||
GenBank mRNA | AF479659.1 | - | 3 | 441 |