Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012057.1 | NP_001012057.1 | 4 | 590 | ||
XM_008772405.1 | XP_008770627.1 | 5 | 711 | |||
XM_008772406.1 | XP_008770628.1 | 5 | 900 | |||
XM_008772407.1 | XP_008770629.1 | 5 | 637 | |||
GenBank mRNA | FQ223845.1 | - | 786 |