Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012084.1 | NP_001012084.1 | 9 | 1703 | ||
XM_006233372.2 | XP_006233434.1 | 9 | 2497 | |||
XM_006233374.1 | XP_006233436.1 | 10 | 1775 | |||
XM_006233375.2 | XP_006233437.1 | 11 | 1986 | |||
XM_006233377.2 | XP_006233439.1 | 9 | 1945 | |||
XM_006233378.2 | XP_006233440.1 | 10 | 1855 | |||
XM_006233379.2 | XP_006233441.1 | 10 | 1852 | |||
XM_008761506.1 | XP_008759728.1 | 8 | 1817 | |||
XM_008761507.1 | XP_008759729.1 | 10 | 4049 | |||
XM_008761508.1 | XP_008759730.1 | 11 | 3909 | |||
XM_008761509.1 | XP_008759731.1 | 11 | 2401 | |||
XM_008761510.1 | XP_008759732.1 | 11 | 4177 | |||
GenBank mRNA | BC083792.1 | - | 9 | 1703 | 7106499 |