Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012095.1 | NP_001012095.1 | 11 | 1690 | ||
XM_006235853.2 | XP_006235915.1 | 9 | 1288 | |||
XM_006235854.2 | XP_006235916.1 | 9 | 1287 | |||
XM_006235856.2 | XP_006235918.1 | 10 | 1344 | |||
GenBank mRNA | BC083635.1 | - | 11 | 1690 | 7134691 |