Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS2 SIRNA Assays ›
Gene Symbol:
CADPS2
Gene Name:
calcium dependent secretion activator 2
Gene Aliases:
CAPS2
Chromosome Location:
Chr.7: 122318424 - 122886759 on Build GRCh38
UniGene ID:
Hs.649459
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25959
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009571.3
NP_001009571.2
26
3883
NM_001167940.1
NP_001161412.1
28
4018
NM_017954.10
NP_060424.9
28
4006
XM_005250695.4
XP_005250752.1
30
3771
XM_005250696.4
XP_005250753.1
29
3762
XM_005250697.4
XP_005250754.1
29
3762
XM_005250698.4
XP_005250755.1
29
3728
XM_005250699.4
XP_005250756.1
29
3750
XM_005250700.4
XP_005250757.1
28
3747
XM_005250701.4
XP_005250758.1
28
3741
XM_005250702.4
XP_005250759.1
27
3726
XM_005250703.4
XP_005250760.1
29
3651
XM_005250704.4
XP_005250761.1
28
3642
XM_005250705.4
XP_005250762.1
27
3627
XM_005250706.4
XP_005250763.1
27
3627
XM_005250707.4
XP_005250764.1
27
3621
XM_005250708.3
XP_005250765.1
25
3597
XM_011516695.2
XP_011514997.1
28
3608
XM_011516696.2
XP_011514998.1
27
3615
XM_011516697.2
XP_011514999.1
26
3606
XM_017012794.1
XP_016868283.1
27
3732
XM_017012795.1
XP_016868284.1
26
3618
XM_017012796.1
XP_016868285.1
31
3386
XM_017012797.1
XP_016868286.1
30
3437
XM_017012798.1
XP_016868287.1
29
3901
XM_017012799.1
XP_016868288.1
29
3688
XM_017012800.1
XP_016868289.1
27
2779
GenBank mRNA
AB046811.1
-
24
2752
AF401638.1
-
26
3552
AK000768.1
-
11
1061
AK025672.1
-
21
2391
AK054853.1
-
13
1165
AK126518.1
-
27
3485
AL833058.1
-
23
2678
AY264289.1
-
28
3610
BC136601.1
-
26
3582
9020591
BC144278.1
-
26
3582
9052798
BC144279.1
-
28
3717
9052799
KJ534798.1
-
27
3464
KJ534800.1
-
25
3446
KJ534801.1
-
26
3461
KJ534802.1
-
26
3455
KJ534803.1
-
27
3476
KJ534804.1
-
28
3485
KJ534805.1
-
25
3446
KJ535060.1
-
14
1690
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to starvation
protein transport
synaptic vesicle priming
positive regulation of exocytosis
dense core granule exocytosis
lipid binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0