Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012144.1 | NP_001012144.1 | 6, 7 | 976 | ||
GenBank mRNA | AB218900.1 | - | 5, 6 | 717 | ||
BC078947.1 | - | 6, 7 | 976 | 7109719 |