Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012351.2 | NP_001012351.1 | 4 | 434 | ||
XM_006248869.2 | XP_006248931.1 | 4 | 471 | |||
XM_006248870.2 | XP_006248932.1 | 4 | 471 | |||
GenBank mRNA | BC089824.1 | - | 4 | 436 | 7309116 | |
Y17326.1 | - | 4 | 353 |
Gene Symbol: LOC100360200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727934.4 | XP_002727980.1 | 2 | 365 |