Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012765.4 | NP_001012783.3 | 13 | 2948 | ||
GenBank mRNA | AK147590.1 | - | 3051 | |||
AK147649.1 | - | 3708 | ||||
AK160942.1 | - | 13 | 2948 | |||
BC035550.1 | - | 1086 | ||||
BC090846.1 | - | 13 | 2683 | 30363147 |