Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320772.1 | NP_001307701.1 | 10 | 1115 | ||
NM_017983.6 | NP_060453.3 | 11 | 1198 | |||
NR_135470.1 | 11 | 1334 | ||||
NR_135471.1 | 10 | 1127 | ||||
XM_017024808.1 | XP_016880297.1 | 9 | 3152 | |||
GenBank mRNA | AK000917.1 | - | 10 | 1116 | ||
AK301830.1 | - | 10 | 1061 | |||
AY691424.1 | - | 11 | 1098 | |||
BC039867.2 | - | 11 | 1142 | 6055425 |