Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258008.1 | NP_001244937.1 | 4, 5 | 667 | ||
NM_017984.4 | NP_060454.3 | 4, 5 | 667 | |||
XM_005250479.2 | XP_005250536.1 | 4, 5 | 1098 | |||
XM_005250480.2 | XP_005250537.1 | 4, 5 | 1098 | |||
XM_006716035.3 | XP_006716098.1 | 4, 5 | 1098 | |||
XM_006716036.2 | XP_006716099.1 | 4, 5 | 602 | |||
XM_006716037.3 | XP_006716100.1 | 4, 5 | 1098 | |||
XM_006716038.3 | XP_006716101.1 | 4, 5 | 1098 | |||
XM_006716040.3 | XP_006716103.1 | 4, 5 | 1098 | |||
XM_017012379.1 | XP_016867868.1 | 4, 5 | 1098 | |||
XM_017012380.1 | XP_016867869.1 | 4, 5 | 602 | |||
GenBank mRNA | AK093038.1 | - | 4, 5 | 602 | ||
AK300745.1 | - | 4, 5 | 523 | |||
AK316553.1 | - | 4, 5 | 855 | |||
AL136735.1 | - | 4, 5 | 538 | |||
BC002725.1 | - | 1, 2 | 256 | 3631266 |