Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013132.2 | NP_001013150.1 | 6 | 841 | ||
XM_006252201.2 | XP_006252263.1 | 7 | 1614 | |||
XM_006252202.2 | XP_006252264.1 | 6 | 1494 | |||
GenBank mRNA | BC062801.1 | - | 6 | 844 | 6889991 | |
FQ213407.1 | - | 6 | 775 |