Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013374.1 | NP_001013392.1 | 4 | 490 | ||
GenBank mRNA | AK041656.1 | - | 2 | 2335 | ||
AK142024.1 | - | 459 | ||||
AK158015.1 | - | 4 | 509 | |||
BC046969.1 | - | 4 | 490 | 6313679 |