Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013380.2 | NP_001013398.2 | 5 | 905 | ||
GenBank mRNA | AK050575.1 | - | 5 | 875 | ||
AK140743.1 | - | 5 | 673 | |||
AK147899.1 | - | 5 | 672 | |||
AK152157.1 | - | 5 | 670 | |||
AK161434.1 | - | 5 | 767 | |||
AK161494.1 | - | 5 | 767 | |||
BC058645.1 | - | 5 | 863 | 6405596 |