Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013429.1 | NP_001013447.1 | 2 | 904 | ||
XM_006248058.2 | XP_006248120.1 | 2 | 960 | |||
XM_006248059.2 | XP_006248121.1 | 2 | 608 | |||
GenBank mRNA | BC091387.1 | - | 2 | 904 | 7321417 |