Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018026.3 | NP_060496.2 | 20 | 2401 | ||
XM_011545162.1 | XP_011543464.1 | 20 | 1961 | |||
XM_011545164.2 | XP_011543466.1 | 20 | 3269 | |||
GenBank mRNA | AB033001.1 | - | 13 | 1303 | ||
AK001071.1 | - | 8 | 706 | |||
AK056361.1 | - | 19 | 1908 | |||
AK096644.1 | - | 19 | 2251 | |||
AK293966.1 | - | 9 | 1012 | |||
AY320283.1 | - | 20 | 2441 | |||
BC003173.1 | - | 5 | 370 | |||
BC009936.1 | - | 6 | 476 | |||
BC010096.1 | - | 20 | 2309 | 3502170 | ||
BC042909.1 | - | 9 | 860 | |||
BC052577.1 | - | 20 | 2300 | 4130626 | ||
BC055288.1 | - | 12 | 1105 |