Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014015.1 | NP_001014037.1 | 11 | 2104 | ||
GenBank mRNA | BC087155.1 | - | 11 | 2104 | 7133162 |