Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014038.2 | NP_001014060.2 | 2 | 154 | ||
XM_006232870.2 | XP_006232932.1 | 2 | 157 | |||
XM_006232871.2 | XP_006232933.1 | 2 | 2113 | |||
GenBank mRNA | BC059161.1 | - | 2 | 110 | 6887770 |