Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AATF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012138.3 | NP_036270.1 | 2 | 363 | ||
XM_011524611.1 | XP_011522913.1 | 2 | 347 | |||
GenBank mRNA | AF083208.1 | - | 2 | 280 | ||
AJ249940.2 | - | 2 | 218 | |||
AK057229.1 | - | 2 | 363 | |||
BC000591.2 | - | 2 | 243 | 3346769 |