Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024135.2 | NP_001019306.2 | 1 | 2279 | ||
GenBank mRNA | AK135854.1 | - | 2 | 1605 | ||
BC096019.1 | - | 1 | 2279 | 6830769 |