Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024269.1 | NP_001019440.1 | 5, 6 | 846 | ||
XM_006246006.2 | XP_006246068.1 | 6, 7 | 811 | |||
XM_006246007.1 | XP_006246069.1 | 6, 7 | 2215 | |||
XM_008767588.1 | XP_008765810.1 | 6, 7 | 1674 | |||
GenBank mRNA | BC089960.1 | - | 5, 6 | 846 | 7313914 |