Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xirp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024618.2 | NP_001019789.1 | 7 | 2802 | ||
NM_001083919.1 | NP_001077388.1 | 7 | 2802 | |||
GenBank mRNA | AY775570.1 | - | 6 | 2656 | ||
AY775571.1 | - | 1 | 1385 | |||
DQ011666.1 | - | 7 | 2803 | |||
EF119716.1 | - | 7 | 2802 | |||
EF122140.1 | - | 6 | 2557 |