Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsku | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024619.3 | NP_001019790.1 | 2 | 549 | ||
NM_001168539.1 | NP_001162011.1 | 2 | 553 | |||
NM_001168540.1 | NP_001162012.1 | 3 | 659 | |||
NM_001168541.1 | NP_001162013.1 | 4 | 799 | |||
GenBank mRNA | AB214184.1 | - | 1 | 494 | ||
AK035461.1 | - | 2 | 550 | |||
AK053973.1 | - | 4 | 800 | |||
AK137208.1 | - | 2 | 550 | |||
BC116310.1 | - | 2 | 561 | 40095310 | ||
BC116311.1 | - | 3 | 671 | 40095311 | ||
BC129977.1 | - | 2 | 561 | 40095312 | ||
BC129978.1 | - | 3 | 671 | 40095313 |