Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134479.1 | NP_001127951.1 | 3 | 1099 | ||
NM_018103.4 | NP_060573.2 | 3 | 1086 | |||
XM_011541689.1 | XP_011539991.1 | 4 | 1170 | |||
XM_017001599.1 | XP_016857088.1 | 3 | 1441 | |||
XM_017001600.1 | XP_016857089.1 | 3 | 896 | |||
XM_017001601.1 | XP_016857090.1 | 3 | 1787 | |||
XM_017001602.1 | XP_016857091.1 | 3 | 1140 | |||
GenBank mRNA | AK001332.1 | - | 1 | 152 | ||
AK092238.1 | - | 3 | 1062 | |||
AK222787.1 | - | 1 | 113 | |||
AL831880.1 | - | 1 | 558 | |||
AL832952.1 | - | 1 | 517 | |||
BC009486.1 | - | 3 | 1031 | |||
BC024159.2 | - | 3 | 1053 | 3958069 |