Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025118.1 | NP_001020289.1 | 1 | 440 | ||
XM_006232886.2 | XP_006232948.1 | 4 | 1664 | |||
XM_008761304.1 | XP_008759526.1 | 5 | 1622 | |||
GenBank mRNA | BC091386.1 | - | 1 | 440 | 7321358 |