Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001624.3 | NP_001615.2 | 17 | 5158 | ||
XM_005266839.3 | XP_005266896.1 | 19 | 5996 | |||
GenBank mRNA | AK025693.1 | - | 6 | 532 | ||
AK297008.1 | - | 7 | 718 | |||
AK300443.1 | - | 11 | 1193 | |||
AK316287.1 | - | 11 | 1194 | |||
CV573838.1 | - | 6 | 570 | |||
U83115.1 | - | 17 | 4413 |