Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009338.3 | NP_033364.2 | 9 | 1763 | ||
GenBank mRNA | AK037249.1 | - | 9 | 1750 | ||
AK151751.1 | - | 9 | 1751 | |||
AK166813.1 | - | 9 | 1774 | |||
AK167234.1 | - | 9 | 1763 | |||
BC004823.1 | - | 9 | 1724 | 2598868 | ||
BC012496.1 | - | 9 | 1739 | 4500929 | ||
M35797.1 | - | 6 | 1344 |