Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170744.1 | NP_001164215.1 | 9 | 3073 | ||
NM_009980.4 | NP_034110.1 | 11 | 1950 | |||
GenBank mRNA | AB033123.1 | - | 11 | 1815 | ||
AF016508.1 | - | 9 | 1422 | |||
AK134399.1 | - | 10 | 1474 | |||
AK146713.1 | - | 11 | 1835 | |||
AK147871.1 | - | 11 | 1833 | |||
AK198744.1 | - | 1 | 235 | |||
AK201071.1 | - | 1 | 235 | |||
AK217772.1 | - | 1 | 237 | |||
BC017520.1 | - | 10 | 2918 | 4511826 |